missing translation for 'onlineSavingsMsg'
Learn More

Nicardipine hydrochloride, 98%

Product Code. 15463879
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463879 5 g 5g
15453879 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463879 Supplier Thermo Scientific Alfa Aesar Supplier No. J61269.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L-type calcium channel blocker | CAS: 54527-84-3 | C26H30ClN3O6 | 515.991 g/mol

L-type calcium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54527-84-3
Molecular Formula C26H30ClN3O6
Molecular Weight (g/mol) 515.991
MDL Number MFCD00057327
InChI Key AIKVCUNQWYTVTO-UHFFFAOYSA-N
Synonym nicardipine hydrochloride, cardene, nicardipine hcl, perdipine, nicodel, loxen, angioglebil, bionicard, nicardil, perdipina
PubChem CID 41114
IUPAC Name 5-O-[2-[benzyl(methyl)amino]ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
SMILES CC1=C(C(C(=C(N1)C)C(=O)OCCN(C)CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OC.Cl

Specifications

Color Yellow
Quantity 5 g
UN Number UN2811
Merck Index 14,6495
Formula Weight 515.99
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nicardipine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.