Learn More
Invitrogen™ NIBP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110514
Description
Immunogen sequence: SHVKTLEAVL NFKYSGGPGH TEGYYRNLSL GLHVEVEPSV FFTRVSTLPA TSTRQCHLLL DVFNSTEHEL TVSTRSSEAL ILHAGECQRM AIQVDKFNFE SFPESPGEKG QFANPKQLEE ERREARGLEI HSKLGICWRI PSLKRSGEAS VEGLLNQLVL EHLQLAPLQW DVLVDGQPCD REAVAACQVG DPVRLEVRLT NRSPRSVGPF ALTVVPFQDH QNGVHNYDLH DTVSFVGSST FYLDAVQPSG QSACLGALLF LYTGDFFLHI RFHEDSTSKE LPPSWFCLPS VHVCALEAQA.
NIBP is a 1246 amino acid intracellular regulatory protein. Reports suggest that NIBP physically interacts with NIK, IKK (beta), but not IKK (alpha) or IKK (gamma) and thus acts as a novel enhancer of cytokine-induced NF-(kappa) B signaling pathway. Due to its neuronal expression NIBP is considered as a potential target for modulating the NF-(kappa) B signaling cascade in neuronal pathologies dependent upon abnormal activation of this pathway. NIBP is mainly expressed in brain, muscle, heart, and kidney.
Specifications
| NIBP | |
| Polyclonal | |
| Unconjugated | |
| TRAPPC9 | |
| 1810044A24Rik; 2900005P22Rik; 4632408O18Rik; hypothetical protein LOC533451; Ibp; IKBKBBP; IKK2 binding protein; Kiaa1882; mKIAA1882; MRT13; Nibp; NIK- and IKBKB-binding protein; NIK and IKK(beta) binding protein; NIK and IKK{beta} binding protein; NIK and IKK-beta binding protein; RGD1309461; T1; trafficking protein particle complex 9; trafficking protein particle complex subunit 9; TRAPP 120 kDa subunit; TRAPPC9; TRS120; TRS130; tularik gene 1 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 76510, 83696 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.44 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q3U0M1, Q96Q05 | |
| TRAPPC9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 947-1246 of human TRAPPC9 (NP_113654.4). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.