missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Niban-like protein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554371
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Immunogen sequence: EVTDMNLNVI NEGGIDKLGE YMEKLSRLAY HPLKMQSCYE KMESLRLDGL QQRFDVSSTS VFKQRAQIHM REQMDNAVYT FETLLHQELG KGPTKEELCK SIQRV Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
FAM129B (Family With Sequence Similarity 129 Member B, Niban Like Protein 1) is a Protein Coding gene. May play a role in apoptosis suppression. May promote melanoma cell invasion in vitro. Among its related pathways are MAP Kinase Signaling. An important paralog of this gene is FAM129A.
Tekniske data
| Niban-like protein | |
| Polyclonal | |
| Unconjugated | |
| Fam129b | |
| 9130404D14Rik; bA356B19.6; C9orf88; FAM129B; family with sequence similarity 129 member B; family with sequence similarity 129, member B; Meg-3; Melanoma invasion by ERK; MINERVA; NIBAN2; Niban-like protein 1; OC58; Protein FAM129B; Protein Niban 2; RGD1307018 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64855 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96TA1 | |
| Fam129b | |
| Recombinant protein corresponding to Human Niban-like protein. Recombinant protein control fragment (Product #RP-92961). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion