missing translation for 'onlineSavingsMsg'
Learn More

Niacin, MedChemExpress

Product Code. 16472059
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472059 1 g 1g
16440788 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472059 Supplier Medchem Express Supplier No. HYB0143
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Niacin (Vitamin B3) is an orally active water-soluble B3 vitamin that is an essential nutrient for humans. Niacin (Vitamin B3) plays a key role in energy metabolism, cell signaling cascades regulating gene expression and apoptosis. Niacin (Vitamin B3) is also used in the study of cardiovascular diseases.

Niacin (Vitamin B3) is an orally active water-soluble B3 vitamin that is an essential nutrient for humans. Niacin (Vitamin B3) plays a key role in energy metabolism, cell signaling cascades regulating gene expression and apoptosis. Niacin (Vitamin B3) is also used in the study of cardiovascular diseases.

Specifications

Chemical Name or Material Niacin
CAS 59-67-6
Color White
Purity Grade Notes Research
Molecular Formula C6H5NO2
Quantity 1 g
Synonym Nicotinic acid Vitamin B3
Solubility Information DMSO : 50 mg/mL (406.14 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 10 mg/mL (81.23 mM; Need ultrasonic)
SMILES O=C(C1=CC=CN=C1)O
Molecular Weight (g/mol) 123.11
Formula Weight 123.11
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.