missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Ni-NTA Purification System with Antibody, with Anti-His (C-term)-HRP Antibody

Product Code. 10083192
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10083192 1 Kit 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10083192 Supplier Novex™ Supplier No. K95301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Ni-NTA Purification System with Antibody, with Anti-His (C-term)-HRP Antibody

The Ni-NTA Purification System is designed for the purification of recombinant proteins that contain a polyhistidine (6xHis) sequence. The kit utilizes Ni-NTA nickel-chelating resin and is supplied with native and denaturing buffers for efficient purification of recombinant proteins under different conditions.
TRUSTED_SUSTAINABILITY

Specifications

Description Ni-NTA Purification System with Antibody, with Anti-His (C-term)-HRP Antibody, 1Kit, 6 Months Shelf Life, 4°C Storage Temperature Ten Milliliters of Ni-NTA Agarose
Product Type Ni-NTA Purification System with Antibody, with Anti-His (C-term)-HRP Antibody
Quantity 1 Kit
Content And Storage 4°C
Column Type Affinity Column
For Use With (Application) Protein purification and isolation

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.