missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NHS-LC-Biotin

Product Code. 18705976
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18705976 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18705976 Supplier Tocris Bioscience™ Supplier No. 7301/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Amine reactive biotinylation reagent, cell permeable

NHS-LC-Biotin is a membrane permeable amine reactive biotinylation reagent used to attach biotin to primary amines with a 6-atom chain (22.4 angstroms) linker. NHS-LC-Biotin is commonly used to label proteins and antibodies. Reactive in pH 7-9.

This product should be dissolved in DMSO (solubility = 100 mM) or DMF prior to further dilution in aqueous buffers.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-[[5-[(3 AS,4 S,6 aR)-Hexahydro-2-oxo-1 H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]hexanoic acid 2,5-dioxo-1-pyrrolidinyl ester
CAS 72040-63-2
Quantity 50 mg
Target Buffers, Chelators and Reagents
Molecular Formula C20H30N4O6S
Purity 0.9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.