missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93118-0.1ml
This item is not returnable.
View return policy
Description
NHP2 Polyclonal antibody specifically detects NHP2 in Human, Rat samples. It is validated for Western Blot
Specifications
| NHP2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ20479, H/ACA ribonucleoprotein complex subunit 2, NHP2 ribonucleoprotein homolog (yeast), NHP2-like protein, NHP2P, NOLA2member 2 (H/ACA small nucleolar RNPs), Nucleolar protein family A member 2, snoRNP protein NHP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-153 of human NHP2 (NP_060308.1). MTKIKADPDGPEAQAEACSGERTYQELLVNQNPIAQPLASRRLTRKLYKCIKKAVKQKQIRRGVKEVQKFVNKGEKGIMVLAGDTLPIEVYCHLPVMCEDRNLPYVYIPSKTDLGAAAGSKRPTCVIMVKPHEEYQEAYDECLEEVQSLPLPL | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 55651 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction