missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems NHP XL IL-5, Magnetic Luminex™ Performance Assay
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Primate |
| Binding Target | IL-5 |
| Gene ID (Entrez) | 3567 |
| Synonym | BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction