missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Immunoassay Reagents and Kits
- Singleplex and Multiplex Bead Based Immunoassays
- R&D Systems NHP XL Cytokine Luminex™ Performance Premixed Kit
R&D Systems NHP XL Cytokine Luminex™ Performance Premixed Kit
Product Code. 18411410
Change view
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
23 Plex
24 Plex
25 Plex
26 Plex
27 Plex
28 Plex
29 Plex
3 Plex
30 Plex
31 Plex
32 Plex
33 Plex
34 Plex
35 Plex
36 Plex
37 Plex
38 Plex
39 Plex
4 Plex
40 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Each
39 product options available for selection
| Product Code. | Quantity | unitSize |
|---|---|---|
| 18411410 | 8 Plex | 1 set |
| 30002208 | 36 Plex | Each |
| 18717479 | 35 Plex | Each |
| 18792298 | 34 Plex | Each |
| 30002207 | 33 Plex | Each |
| 30002206 | 32 Plex | Each |
| 30002205 | 31 Plex | Each |
| 30002204 | 30 Plex | Each |
| 30002203 | 29 Plex | Each |
| 18700609 | 28 Plex | Each |
| 18724969 | 27 Plex | Each |
| 30002202 | 26 Plex | Each |
| 30002201 | 25 Plex | Each |
| 30002200 | 24 Plex | Each |
| 30002199 | 23 Plex | Each |
| 18742129 | 22 Plex | Each |
| 30002198 | 21 Plex | Each |
| 30002197 | 20 Plex | Each |
| 30002196 | 19 Plex | Each |
| 30002195 | 18 Plex | Each |
| 30002194 | 17 Plex | Each |
| 30002193 | 16 Plex | Each |
| 30002192 | 15 Plex | Each |
| 30002191 | 14 Plex | Each |
| 18703619 | 13 Plex | Each |
| 18729578 | 12 Plex | Each |
| 30002190 | 11 Plex | Each |
| 30002189 | 10 Plex | Each |
| 18795189 | 9 Plex | Each |
| 30002188 | 7 Plex | Each |
| 30002187 | 6 Plex | Each |
| 18707479 | 5 Plex | Each |
| 18709787 | 4 Plex | Each |
| 30002186 | 3 Plex | Each |
| 30002185 | 2 Plex | Each |
| 18441820 | 40 Plex | 1 set |
| 18451820 | 38 Plex | 1 set |
| 18422530 | 39 Plex | 1 set |
| 18452080 | 37 Plex | 1 set |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 39 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 8 Plex |
| Species | Primate |
| Binding Target | Multiplex Cytokine Assay Kits |
| For Use With (Application) | ELISA |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?