missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems NHP XL Cytokine Luminex™ Performance Premixed Kit

Product Code. 18452080
Change view
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
23 Plex
24 Plex
25 Plex
26 Plex
27 Plex
28 Plex
29 Plex
3 Plex
30 Plex
31 Plex
32 Plex
33 Plex
34 Plex
35 Plex
36 Plex
37 Plex
38 Plex
39 Plex
4 Plex
40 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Each
39 product options available for selection
Product selection table with 39 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18452080 37 Plex 1 set
30002208 36 Plex Each
18717479 35 Plex Each
18792298 34 Plex Each
30002207 33 Plex Each
30002206 32 Plex Each
30002205 31 Plex Each
30002204 30 Plex Each
30002203 29 Plex Each
18700609 28 Plex Each
18724969 27 Plex Each
30002202 26 Plex Each
30002201 25 Plex Each
30002200 24 Plex Each
30002199 23 Plex Each
18742129 22 Plex Each
30002198 21 Plex Each
30002197 20 Plex Each
30002196 19 Plex Each
30002195 18 Plex Each
30002194 17 Plex Each
30002193 16 Plex Each
30002192 15 Plex Each
30002191 14 Plex Each
18703619 13 Plex Each
18729578 12 Plex Each
30002190 11 Plex Each
30002189 10 Plex Each
18795189 9 Plex Each
30002188 7 Plex Each
30002187 6 Plex Each
18707479 5 Plex Each
18709787 4 Plex Each
30002186 3 Plex Each
30002185 2 Plex Each
18441820 40 Plex 1 set
18451820 38 Plex 1 set
18422530 39 Plex 1 set
18411410 8 Plex 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 39 options available.
39 options
This item is not returnable. View return policy
Product Code. 18452080 Supplier R&D Systems Supplier No. FCSTM2137

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 37 Plex
Species Primate
Binding Target Multiplex Cytokine Assay Kits
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.