missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NHERF2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518436
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
NHE3 is a sodium/hydrogen exchanger in the brush border membrane of the proximal tubule, small intestine, and colon that plays a major role in transepithelial sodium absorption. SLC9A3R2, as well as SLC9A3R1 and protein kinase A phosphorylation, may play a role in NHE3 regulation.
Specifications
| NHERF2 | |
| Polyclonal | |
| Unconjugated | |
| SLC9A3R2 | |
| 0610011L07Rik; 1200011K07Rik; 2010007A20Rik; E3karp; HGNC:11076; MGC104639; Na(+)/H(+) exchange regulatory cofactor NHE-RF2; NHE3 kinase A regulatory protein E3KARP; NHE3 regulatory factor 2; NHE3RF2; Nherf2; NHERF-2; OCTS2; SIP1; SIP-1; SLC9A3 regulator 2; SLC9A3R2; sodium/hydrogen exchanger; sodium/hydrogen exchanger 3 kinase A regulatory protein; Sodium-hydrogen exchanger regulatory factor 2; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 2; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatory factor 2; solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2; solute carrier family 9 isoform 3 regulator 2; solute carrier family 9 isoform 3 regulator 2, isoform A; sodium/hydrogen exchanger; solute carrier family 9 isoform A3 regulatory factor 2; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 2; Sry interacting protein 1; SRY-interacting protein 1; Sryip1; TKA-1; tyrosine kinase activator protein 1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 9351 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q15599 | |
| SLC9A3R2 | |
| Synthetic peptide sequence (DGSAWKQDPFQ) corresponding to the internal amino acids of SLC9A3R2 (aa 286-297). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction