missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHERF-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£305.00 - £435.00
Specifications
| Antigen | NHERF-1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18440601
|
Novus Biologicals
NBP1-84919-25ul |
25ul |
£305.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18286927
|
Novus Biologicals
NBP1-84919 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NHERF-1 Polyclonal specifically detects NHERF-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NHERF-1 | |
| Polyclonal | |
| Rabbit | |
| Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 9368 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GENVEKETHQQVVSRIRAALNAVRLLVVDPETDEQLQKLGVQVREELLRAQEAPGQAEPPAAAEVQGAGNENEPREADKSHPEQRELRPRLCTMKKGPSGYGFNLHSD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| EBP50NHERF-1, ezrin-radixin-moesin binding phosphoprotein-50, Ezrin-radixin-moesin-binding phosphoprotein 50, Na(+)/H(+) exchange regulatory cofactor NHE-RF1, Na+/H+ exchange regulatory co-factor, NHERF1solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 1, NHERFNPHLOP2, Regulatory cofactor of Na(+)/H(+) exchanger, sodium/hydrogen exchanger regulatory factor 1, Sodium-hydrogen exchanger regulatory factor 1, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatoryfactor 1, solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 1, Solute carrier family 9 isoform A3 regulatory factor 1 | |
| SLC9A3R1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title