missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE8 Antibody (7A11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-62091H
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
NHE8 Monoclonal specifically detects NHE8 in Human, Mouse, Rat samples. It is validated for Western Blot.
Tekniske data
| NHE8 | |
| Monoclonal | |
| HRP | |
| Q9Y2E8 | |
| SLC9A8 | |
| A fusion protein containing the C-terminal 89 residues of human NHE8. [Swiss-Prot# Q9Y2E8] | |
| Protein G purified | |
| RUO | |
| Primary | |
| This is specific for the 85 kDa mature glycosylated form of the protein. It does not recognize the 55 kDa immature form. | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| 7A11 | |
| Western Blot | |
| DKFZp686C03237, KIAA0939DKFZp686C03237, MGC138418, Na(+)/H(+) exchanger 8, NHE-8, NHE8FLJ42500, sodium/hydrogen exchanger 8, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 8, solute carrier family 9 (sodium/hydrogen exchanger), member 8, Solute carrier family 9 member 8 | |
| Mouse | |
| 80 kDa | |
| 0.1 mL | |
| Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism | |
| 23315 | |
| Human, Mouse, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion