missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE8 Antibody (7A11), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-62091AF532
This item is not returnable.
View return policy
Description
NHE8 Monoclonal specifically detects NHE8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| NHE8 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SLC9A8 | |
| A fusion protein containing the C-terminal 89 residues of human NHE8. [Swiss-ProtNo. Q9Y2E8] | |
| 0.1 mL | |
| Primary | |
| This is specific for the 85 kDa mature glycosylated form of the protein. It does not recognize the 55 kDa immature form. | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 7A11 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| DKFZp686C03237, KIAA0939DKFZp686C03237, MGC138418, Na(+)/H(+) exchanger 8, NHE-8, NHE8FLJ42500, sodium/hydrogen exchanger 8, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 8, solute carrier family 9 (sodium/hydrogen exchanger), member 8, Solute carrier family 9 member 8 | |
| Mouse | |
| Protein G purified | |
| Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism | |
| 23315.0 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction