missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE3/SLC9A3, p Ser552 Antibody (14D5) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80877
This item is not returnable.
View return policy
Description
NHE3/SLC9A3 (p Ser552) Monoclonal antibody specifically detects NHE3/SLC9A3 (p Ser552) in Human, Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoassay, Knockdown.
Specifications
| NHE3/SLC9A3 (p Ser552) | |
| Monoclonal | |
| 2 mg/mL | |
| Western Blot : 1-5 ug/ml, Simple Western : 1:1000, Flow Cytometry reported in scientific literature (PMID 21255452), ELISA : 1:100-1:2000, Immunohistochemistry : 1:50-1:1000, Immunocytochemistry/Immunofluorescence : 1:50-1:1000, Immunoprecipitation 1:10-1:500, Knockout Validated reported in scientific literature (PMID 31276916), Knockdown Validated reported in scientific literature (PMID 31276916) | |
| isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Porcine reactivity reported in scientific literature (PMID: 29510506). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoassay, KnockDown | |
| 14D5 | |
| Unconjugated | |
| Tris-Glycine, 0.15 M NaCl | |
| SLC9A3 | |
| Synthetic peptide made to a region surrounding the Serine 552 portion of rat NHE3. [UniProt# P26433] | |
| 0.1 mL | |
| Lipid and Metabolism, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 6550 | |
| Human, Mouse, Rat, Pig, Opossum, Primate, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction