missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE3/SLC9A3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£166.00 - £368.00
Specifications
| Antigen | NHE3/SLC9A3 |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18380293
|
Novus Biologicals
NBP3-00589 |
0.1 mg |
£368.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603057
|
Novus Biologicals
NBP3-00589SS |
0.025 mg |
£166.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NHE3/SLC9A3 Polyclonal antibody specifically detects NHE3/SLC9A3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| NHE3/SLC9A3 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3 | |
| Partial synthetic peptide made to an internal portion of the human NHE3 protein (between amino acids 600-660) [UniProt P48764] | |
| Immunogen affinity purified |
| 1.0 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| PBS | |
| 6550 | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title