missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NHE3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583345
This item is not returnable.
View return policy
Description
Immunogen sequence: DNPVFSPDEA LDRSLLARLP PWLSPGETVV PSQRARTQIP YSPGTFCRLM PFRLSSKSVD SFLQADGPEE RPPAALPEST Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 79%.
NHE3 (also known as SLC9A3 - Sodium/hydrogen exchanger 3) is a member of the sodium/hydrogen exchanger family that plays a crucial role in acid-base and volume homeostasis by mediating the majority of sodium and bicarbonate reabsorption in the proximal tubule of the kidney. Mutations in the gene can lead to Diarrhea 8.
Specifications
| NHE3 | |
| Polyclonal | |
| Unconjugated | |
| SLC9A3 | |
| 9030624O13Rik; AI930210; DIAR8; MGC126718; MGC126720; Na(+)/H(+) exchanger 3; Na/H exchanger 3; Na+/H+ exchanger homolog; Na+/H+ exchanger isoform 3; membrane ion exchange protein; Nhe3; NHE-3; NHE-3 antibody; plasma membrane ion transport protein; putative sodium hydrogen exchanger subtype 3; Slc9a3; SLC9A3 antibody; sodium proton exchanger; sodium/hydrogen exchanger; sodium/hydrogen exchanger 3; sodium/hydrogen exchanger isoform 3; Solute carrier family 9 (sodium/hydrogen exchanger 3), antiporter 3, Na+/H+ (amiloride insensitive); solute carrier family 9 (sodium/hydrogen exchanger), isoform 3; solute carrier family 9 (sodium/hydrogen exchanger), member 3; solute carrier family 9 member 3; solute carrier family 9 member A3; solute carrier family 9, member 3; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6550 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P48764 | |
| SLC9A3 | |
| Recombinant protein corresponding to Human SLC9A3. Recombinant protein control fragment (Product #RP-96810). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction