missing translation for 'onlineSavingsMsg'
Learn More

NHE1/SLC9A1 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18411714 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411714 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411714 Supplier Novus Biologicals Supplier No. NBP176847UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NHE1/SLC9A1 Polyclonal antibody specifically detects NHE1/SLC9A1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NHE1/SLC9A1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias APNH1, APNHFLJ42224, Na(+)/H(+) antiporter, amiloride-sensitive, Na(+)/H(+) exchanger 1, Na+/H+, amiloride sensitive), Na-Li countertransporter, NHE-1, NHE1Na+/H+ antiporter, amiloride-sensitive, sodium/hydrogen exchanger 1, solute carrier family 9 (sodium/hydrogen exchanger), isoform 1 (antiporter, solute carrier family 9 (sodium/hydrogen exchanger), member 1, solute carrier family 9 (sodium/hydrogen exchanger), member 1 (antiporter, Solute carrier family 9 member 1
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of the human Nhe-1. The immunogen is located within the last 50 amino acids of Nhe-1. Amino Acid Squence: CLSDPGPHPEPGEGEPFFPK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6548
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.