Learn More
NHE1 Mouse anti-Human, Unconjugated, Clone: 3D9F3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67363-1-IG-150UL
This item is not returnable.
View return policy
Description
The NHE1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NHE1. This antibody recognizes human, rat antigen. The NHE1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| NHE1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat |
| Western Blot, Immunohistochemistry, ELISA | |
| 3D9F3 | |
| Unconjugated | |
| 150 μL | |
| 6548 | |
| IgG1 |