missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHA2/SLC9B2/NHEDC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59579
This item is not returnable.
View return policy
Description
NHA2/SLC9B2/NHEDC2 Polyclonal specifically detects NHA2/SLC9B2/NHEDC2 in Human samples. It is validated for Western Blot.
Specifications
| NHA2/SLC9B2/NHEDC2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ23984, Mitochondrial Na(+)/H(+) exchanger NHA2, mitochondrial sodium/hydrogen exchanger NHA2, Na(+)/H(+) exchanger-like domain-containing protein 2, Na+/H+ exchanger domain containing 2, NHA2NHE10, NHE domain-containing protein 2, Sodium/hydrogen exchanger-like domain-containing protein 2 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 133308 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q86UD5 | |
| SLC9B2 | |
| Synthetic peptides corresponding to NHEDC2(Na+/H+ exchanger domain containing 2) The peptide sequence was selected from the C terminal of NHEDC2. Peptide sequence IFISFAWLPKATVQAAIGSVALDTARSHGEKQLEDYGMDVLTVAFLSILI. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Rat: 100%; Mouse: 100%; Sumatran orangutan: 100%; Human: 100%; Bovine: 92%; Canine: 85%;. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction