missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHA2/SLC9B2/NHEDC2 Antibody (896151) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8404W
This item is not returnable.
View return policy
Description
NHA2/SLC9B2/NHEDC2 Monoclonal antibody specifically detects NHA2/SLC9B2/NHEDC2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| NHA2/SLC9B2/NHEDC2 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 133308 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 896151 | |
| Flow Cytometry, CyTOF-ready | |
| FLJ23984, Mitochondrial Na(+)/H(+) exchanger NHA2, mitochondrial sodium/hydrogen exchanger NHA2, Na(+)/H(+) exchanger-like domain-containing protein 2, Na+/H+ exchanger domain containing 2, NHA2NHE10, NHE domain-containing protein 2, Sodium/hydrogen exchanger-like domain-containing protein 2 | |
| NS0 mouse myeloma cell line transfected with human NHA2/SLC9B2, Met1-Val537, Accession # NP_849155 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction