missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGRN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14885-1-AP-150UL
This item is not returnable.
View return policy
Description
NGRN may be involved in neuronal differentiation.Specifications
| NGRN | |
| Polyclonal | |
| Unconjugated | |
| NGRN | |
| DSC92, FI58G, Neugrin, NGRN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 499191, 51335, 83485 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q3T1H2, Q99KS2, Q9NPE2 | |
| NGRN | |
| NGRN Fusion Protein Ag6661 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction