missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NgR3/NgRH2/RTN4RL1 Antibody (S03-6C3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19895-50ul
This item is not returnable.
View return policy
Description
NgR3/NgRH2/RTN4RL1 Monoclonal antibody specifically detects NgR3/NgRH2/RTN4RL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| NgR3/NgRH2/RTN4RL1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 146760 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| S03-6C3 | |
| Unconjugated | |
| DKFZp547J144, NgR3, NGRH2, NGRL2, nogo receptor-like 2, Nogo-66 receptor homolog 2, Nogo-66 receptor-related protein 3, reticulon 4 receptor-like 1, reticulon-4 receptor-like 1 | |
| A synthetic peptide of human NgR3/NgRH2/RTN4RL1 (Uniprot # Q86UN2) | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction