missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NgR2/NgRH1/RTN4RL2 Antibody (P14), Janelia Fluor™ 549, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52676JF549
This item is not returnable.
View return policy
Description
NgR2/NgRH1/RTN4RL2 Monoclonal specifically detects NgR2/NgRH1/RTN4RL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| NgR2/NgRH1/RTN4RL2 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RTN4RL2 | |
| (His)6-tagged fragment of rat NgR2 (amino acids 279-420). | |
| 0.1 mL | |
| 349667 | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| P14 | |
| Flow Cytometry | |
| NgR2, NGRH1, reticulon 4 receptor-like 2 | |
| Rabbit | |
| Protein A or G purified | |
| Primary | |
| The antibody binds to human NgR2 with a Kd of 2.3 nM and to rat NgR2 with a Kd of 0.48 nM (bivalent binding). | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction