missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NgR2/NgRH1/RTN4RL2 Antibody (P14), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52676DL594
This item is not returnable.
View return policy
Description
NgR2/NgRH1/RTN4RL2 Monoclonal antibody specifically detects NgR2/NgRH1/RTN4RL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF
Specifications
| NgR2/NgRH1/RTN4RL2 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF | |
| P14 | |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF-ready | |
| NgR2, NGRH1, reticulon 4 receptor-like 2 | |
| (His)6-tagged fragment of rat NgR2 (amino acids 279-420). | |
| 0.1 mL | |
| Cellular Signaling | |
| 349667 | |
| Store at 4C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction