missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NgR2/NgRH1/RTN4RL2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41200-0.025mg
This item is not returnable.
View return policy
Description
NgR2/NgRH1/RTN4RL2 Polyclonal specifically detects NgR2/NgRH1/RTN4RL2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| NgR2/NgRH1/RTN4RL2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| RTN4RL2 | |
| Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human RTN4RL2. The immunogen is located within amino acids 330 - 380 of RTN4RL2. Amino Acid Squence: LYRDLPAEDSRGRQGGDA | |
| Affinity Purified | |
| RUO | |
| 349667 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA | |
| NgR2, NGRH1, reticulon 4 receptor-like 2 | |
| Rabbit | |
| 46 kDa | |
| 0.025 mg | |
| Primary | |
| Three alternatively spliced transcript variants have been observed. RTN4RL2 antibody is predicted to not cross-react with other Nogo receptor-like proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction