missing translation for 'onlineSavingsMsg'
Learn More

NGP555, MedChemExpress

Product Code. 16455176
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455176 100 mg 100mg
16433006 25 mg 25mg
16426957 5 mg 5mg
16414238 10 mg 10mg
16410577 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16455176 Supplier Medchem Express Supplier No. HY108714

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NGP555 is a γ-secretase modulator.

NGP555 is a γ-secretase modulator.

Specifications

Chemical Name or Material NGP555
CAS 1304630-27-0
Color White
Purity Grade Notes Research
Molecular Formula C23H23FN4S
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (61.50 mM; Need ultrasonic)
SMILES CC1=CN(C2=CC=C(C3=CSC(NC4=CC(CC)=C(C)C=C4C)=N3)C=C2F)C=N1
Molecular Weight (g/mol) 406.52
Formula Weight 406.52
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.