missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford NGM
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
Description
A standard formulation for use in Nematode growth studies.
A standard formulation for use in Nematode growth studies.
Contains:
Agar 17.5 g/l
Sodium Chloride 3.0 g/l
Peptone 2.5 g/l
Cholesterol 0.005 g/l
Specifications
Specifications
| Quantity | 1 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction