missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFRAP1, Mouse anti-Human, Clone: 4E5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00027018-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MANIHQENEEMEQPMQNGEEDRPLGGGEGHQPAGNRRGQARRLAPNFRWAIPNRQINDGMGGDGDDMEIFMEEMREIRRKLRELQLRNCLRILMGELSNHHDHHDEFCLMSpecifications
| NGFRAP1 | |
| Monoclonal | |
| Unconjugated | |
| nerve growth factor receptor (TNFRSF16) associated protein 1 | |
| BEX3/Bex/DXS6984E/HGR74/NADE | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4E5 | |
| In 1x PBS, pH 7.4 | |
| BC003190 | |
| NGFRAP1 | |
| NGFRAP1 (AAH03190, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 27018 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction