missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFRAP1/BEX3/NADE Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31652
This item is not returnable.
View return policy
Description
NGFRAP1/BEX3/NADE Polyclonal specifically detects NGFRAP1/BEX3/NADE in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| NGFRAP1/BEX3/NADE | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal | |
| BEX3 | |
| Recombinant protein encompassing a sequence within the center region of human NADE. The exact sequence is proprietary. | |
| 0.1 mL | |
| Apoptosis | |
| 27018 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.38mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| BEX3p75NTR-associated cell death executor, Brain-expressed X-linked protein 3, DXS6984EX-linked 3, HGR74, nerve growth factor receptor (TNFRSF16) associated protein 1, Nerve growth factor receptor-associated protein 1, Ovarian granulosa cell 13.0 kDa protein HGR74, ovarian granulosa cell protein (13kD), protein BEX3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction