missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFR Monoclonal Antibody (NGFR5), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513311
This item is not returnable.
View return policy
Description
MA5-13311 targets Neurotrophin Receptor /NGF-Receptor (p75NGFR) in FACS, WB and ICC/IF applications and shows reactivity with Feline, Ferret, Human, Non-human primate, Rat and Rabbit samples. The MA5-13311 immunogen is nGFR from A875 melanoma cells.
Neurotrophic factors control the survival, differentiation and maintenance of neurons in the peripheral and central nervous systems, and of other neural crest-derived cell types. Developing sympathetic neurons are absolutely dependent upon nerve growth factor (NGF) during the period of target competition in vivo. Antibodies reacting specifically with NGFR p75 are useful tools in the detection and characterization of NGFR p75, to enhance our understanding of a wide range of phenomena in the development, plasticity and repair of the nervous system.
Specifications
| NGFR | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P07174, P08138 | |
| Ngfr | |
| NGFR from A875 melanoma cells | |
| 500 μL | |
| Primary | |
| Feline, Ferret, Human, Non-human Primate, Rabbit, Rat | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| NGFR5 | |
| Biotin | |
| Ngfr | |
| CD271; CD271 antigen; Gp80-LNGFR; LNGFR; low affinity nerve growth factor receptor; low affinity neurotrophin receptor p75NTR; LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 16; low-affinity nerve growth factor receptor; nerve growth factor receptor; nerve growth factor receptor (TNFR superfamily, member 16); nerve growth factor receptor, fast; NGF receptor; NGFR; NGR; p75; p-75; p75 ICD; p75 neurotrophin receptor; p75 ntr; p75(NTR); p75NGFR; p75NTR; RNNGFRR; TNFR superfamily, member 16; Tnfrsf16; TNR16; Tumor necrosis factor receptor superfamily member 16 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100349095, 101101519, 101680416, 24596, 4804 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction