missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFI-B alpha/Nur77/NR4A1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
NGFI-B alpha/Nur77/NR4A1 Polyclonal antibody specifically detects NGFI-B alpha/Nur77/NR4A1 in Human, Mouse samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP)
Specifications
Specifications
| Antigen | NGFI-B alpha/Nur77/NR4A1 |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP) |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Early response protein NAK1, GFRP1ST-59, growth factor-inducible nuclear protein N10, HMRNP10, hormone receptor, MGC9485, N10, NAK1, NAK-1, NGFIB, Nuclear hormone receptor NUR/77, nuclear receptor subfamily 4 group A member 1, nuclear receptor subfamily 4, group A, member 1, NUR77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, steroid receptor TR3, Testicular receptor 3, TR3, TR3 orphan receptor |
| Host Species | Rabbit |
| Immunogen | Partial synthetic peptide made to an internal portion of the human NGFI-B alpha protein (between amino acids 200-300) [UniProt P22736]. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?