missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGEF, p Tyr87 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23339
This item is not returnable.
View return policy
Description
NGEF Polyclonal antibody specifically detects NGEF in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunofluorescence
Specifications
| NGEF | |
| Polyclonal | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:200 | |
| EPHEXIN, ephexin-1, Eph-interacting exchange protein, neuronal guanine nucleotide exchange factorARHGEF27 | |
| Phospho-NGEF (Tyr-87) synthetic peptide (coupled to carrier protein) corresponds to amino acids surrounding tyrosine 87 in mouse NGEF. This sequence has significant homology to the conserved region in human and rat NGEF. [UniProt# Q8CHT1] | |
| 0.1 mL | |
| GPCR, Neuroscience, Signal Transduction | |
| 25791 | |
| Store at -20°C. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction