Learn More
Invitrogen™ NGDN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120276
Description
The target is usually found in the following locations: Cell projection, Chromosome, Cytoplasm, Nucleus, axon, centromere, dendrite, filopodium, nucleolus. Immunogen sequence: LMYLMDLTHL ILDKASGGSL QGHDAVLRLV EIRTVLEKLR PLDQKLKYQI DKLIKTAVTG SLSENDPLRF KPHPSNMMSK LSSEDEEEDE AEDDQSEASG KKSVKGVSKK YVPPRLVPVH YDETEAEREK KRLERAKRRA LSSSVIREL.
Involved in the translational repression of cytoplasmic polyadenylation element (CPE)-containing mRNAs.
Specifications
| NGDN | |
| Polyclonal | |
| Unconjugated | |
| Ngdn | |
| 1500001L15Rik; C14orf120; CANu1; Centromere accumulated nuclear protein 1; EIF4E-binding protein; eukaryotic initiation factor 4E and CPEB binding protein; LCP5; lpd-2; neuroguidin; neuroguidin, EIF4E binding protein; NGD; Ngdn; RGD1310211 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25983, 305887 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8NEJ9 | |
| Ngdn | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 61-209 of human NGDN (NP_001036100.1). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.