missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGDN Antibody (OTI1H4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72979APC
This item is not returnable.
View return policy
Description
NGDN Monoclonal antibody specifically detects NGDN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| NGDN | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 25983 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI1H4 | |
| Western Blot, Immunohistochemistry | |
| C14orf120, DKFZp564O092, EIF4E-binding protein, eukaryotic initiation factor 4E and CPEB binding protein, LCP5, lpd-2, neuroguidin, neuroguidin, EIF4E binding protein, NGDchromosome 14 open reading frame 120 | |
| Full length human recombinant protein of human NGDN (NP_001036100) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction