missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NGB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563594
This item is not returnable.
View return policy
Description
Immunogen sequence: ESLLYMLEKC LGPAFTPATR AAWSQLYGAV VQAMSRGWDG E Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.
Globins are a superfamily of gas-binding heme proteins that are present in bacteria, protists, fungi, plants and animals. Globins play evolutionarily divergent roles which include binding, transport, scavenging, detoxification, and sensing of oxygen, nitric oxide and carbon monoxide. Neuroglobin (Ngb) is a hexacoordinate hemoglobin that is predominantly expressed in the vertebrate brain and may enhance oxygen supply to neural components. Neuroglobin displays a high affinity for oxygen and its presence in cerebral neurons suggests a role in neuronal responses to hypoxia or ischemia. For example, in vitro neuronal hypoxia causes an elevation in the levels of neuroglobin, which enhances neuronal cell survival. The human neuroglobin gene maps to chromosome 14q24 and encodes a 151 amino acid protein.
Specifications
| NGB | |
| Polyclonal | |
| Unconjugated | |
| NGB | |
| Neuroglobin; NGB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 58157 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NPG2 | |
| NGB | |
| Recombinant protein corresponding to Human NGB. Recombinant protein control fragment (Product #RP-104263). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction