Learn More
Invitrogen™ NGAL Monoclonal Antibody (28)
Mouse Monoclonal Antibody
Brand: Invitrogen™ ABS0482802
Description
ABS 048-28-02 binds free porcine NGAL in solution. This antibody does not cross-react with human, dog, mouse, or rat. ABS 048-28-02 can be used as a capture antibody in a sandwich ELISA using biotinylated detection antibody ABS 048-17-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
NGAL (Neutrophil gelatinase-associated lipocalin, lipocalin 2, siderocalin and neutrophil lipocalin) is a 25 kDa glycosylated single chain monomer and member of the lipocalin family of proteins that bind and transport small lipophilic molecules. NGAL is released by activated neutrophils, can form dimers and small amounts of higher oligomers, and complexes with matrix metalloproteinase 9 (MMP-9, gelatinase B). Low level expression of NGAL in a variety of epithelia may be increased in inflammation or cancers. Expression of NGAL in the kidney is dramatically increased by ischemic or nephrotoxic kidney injury.
Specifications
| NGAL | |
| Monoclonal | |
| 1.04 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| 0 | |
| LCN2 | |
| Recombinant pig NGAL adsorbed onto aluminum hydroxide gel. | |
| 400 μL | |
| Primary | |
| Pig | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 28 | |
| Unconjugated | |
| LCN2 | |
| 24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; alpha-2-microglobulin-related protein; Alpha-2U globulin-related protein; AW212229; HGNC:6526; HNL; LCN2; lipocalin 2; lipocalin 2 (oncogene 24p3); Lipocalin 24p3; Lipocalin2; Lipocalin-2; migration-stimulating factor inhibitor; MSFI; neutrophil gelatinase-associated lipocalin; NGAL; oncogene 24p3; OTTHUMP00000022240; p25; RP23-161B9.11; secreted inducible protein 24; Siderocalin; Siderocalin LCN2; Sip24; SV-40-induced 24P3 protein; uterocalin | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100153501 | |
| 4°C, store in dark | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.