missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NGAL Monoclonal Antibody (04)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ ABS0380402

Product Code. 15376281

  • £694.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specificity: ABS 038-04-02 does not cross-react with NGAL from other mammalian species tested. Reactivity: ABS 038-04-02 binds human NGAL in solution and coated onto a solid phase. ABS 038-04-02 can be used as a capture antibody in a sandwich ELISA using biotinylated detection antibodies HYB 211-01-02, ABS 038-14-02, ABS 038-15-02 and ABS 038-23-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

NGAL (Neutrophil gelatinase-associated lipocalin, lipocalin 2, siderocalin and neutrophil lipocalin) is a 25 kDa glycosylated single chain monomer and member of the lipocalin family of proteins that bind and transport small lipophilic molecules. NGAL is released by activated neutrophils, can form dimers and small amounts of higher oligomers, and complexes with matrix metalloproteinase 9 (MMP-9, gelatinase B). Low level expression of NGAL in a variety of epithelia may be increased in inflammation or cancers. Expression of NGAL in the kidney is dramatically increased by ischemic or nephrotoxic kidney injury.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NGAL
Monoclonal
0.99 mg/mL
PBS with 15mM sodium azide; pH 7.4
P80188
LCN2
Recombinant human NGAL adsorbed onto aluminum hydroxide gel.
400 μL
Primary
Human
Antibody
IgG1 κ
ELISA, Western Blot
04
Unconjugated
LCN2
24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; alpha-2-microglobulin-related protein; Alpha-2U globulin-related protein; AW212229; HGNC:6526; HNL; LCN2; lipocalin 2; lipocalin 2 (oncogene 24p3); Lipocalin 24p3; Lipocalin2; Lipocalin-2; migration-stimulating factor inhibitor; MSFI; neutrophil gelatinase-associated lipocalin; NGAL; oncogene 24p3; OTTHUMP00000022240; p25; RP23-161B9.11; secreted inducible protein 24; Siderocalin; Siderocalin LCN2; Sip24; SV-40-induced 24P3 protein; uterocalin
Mouse
Protein A/G
RUO
3934
4°C, store in dark
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.