missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NG-Monomethyl-L-arginine Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | NG-Monomethyl-L-arginine Acetate |
| CAS | 53308-83-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C7 H16 N4 O2 . C2 H4 O2 |
| Quantity | 50 mg |
| Synonym | N5-[Imino(methylamino)methyl]-L-ornithine Acetate, L-NMMA, ANO 1020, Tilarginine Acetate |
| InChI Formula | InChI=1S/C7H16N4O2.C2H4O2/c1-10-7(9)11-4-2-3-5(8)6(12)13;1-2(3)4/h5H,2-4,8H2,1H3,(H,12,13)(H3,9,10,11);1H3,(H,3,4)/t5-;/m0./s1 |
| SMILES | CNC(=N)NCCC[C@H](N)C(=O)O.CC(=O)O |
| IUPAC Name | acetic acid;(2S)-2-amino-5-[(N-methylcarbamimidoyl)amino]pentanoic acid |
| Molecular Weight (g/mol) | 248.28 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?