Learn More
LNI Swissgas NG CASTORE PRO 350
Product Code. 17822259 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage unitSize
17822259 100/115/220/230 V Each
1 options
Product Code. 17822259 Supplier LNI Swissgas Supplier No. 6920.50.350
  • ONLINE EXCLUSIVE PRICE
    £24125.00
    £23695.65 / Pack of 1
    Save £429.35
    1% Off
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Nitrogen generator.
  • Needs source of dry and oil free air.
Specifications
Voltage 100/115/220/230 V
Type Nitrogen Gas Generator