missing translation for 'onlineSavingsMsg'
Learn More

NG 52, MedChemExpress

Product Code. 16448917
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16448917

Brand: Medchem Express HY15154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NG 52 is a potent, cell-permeable, selective, ATP-compatible and orally active Cdc28p and Pho85p kinase inhibitor with IC50s of 7 μM and 2 μM, respectively. NG 52 also inhibits the activity of phosphoglycerate kinase 1 (PGK1) with an IC50 of 2.5 μM. NG 52 is inactive against yeast kinases Kin28p, Srb10, and Cak1p.

NG 52 is a potent, cell-permeable, selective, ATP-compatible and orally active Cdc28p and Pho85p kinase inhibitor with IC50s of 7 μM and 2 μM, respectively. NG 52 also inhibits the activity of phosphoglycerate kinase 1 (PGK1) with an IC50 of 2.5 μM. NG 52 is inactive against yeast kinases Kin28p, Srb10, and Cak1p.

Specifications

Chemical Name or Material NG 52
CAS 212779-48-1
Color White
Purity Grade Notes Research
Molecular Formula C16H19ClN6O
Quantity 5 mg
Synonym Compound 52
Solubility Information DMSO : 100 mg/mL (288.34 mM; Need ultrasonic)
SMILES ClC1=CC(NC2=NC(NCCO)=NC3=C2N=CN3C(C)C)=CC=C1
Molecular Weight (g/mol) 346.81
Formula Weight 346.81
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.