missing translation for 'onlineSavingsMsg'
Learn More

NG 52, MedChemExpress

Product Code. 16422447
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422447 50 mg 50mg
16486328 10 mg 10mg
16466646 100 mg 100mg
16448917 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16422447 Supplier Medchem Express Supplier No. HY15154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NG 52 is a potent, cell-permeable, selective, ATP-compatible and orally active Cdc28p and Pho85p kinase inhibitor with IC50s of 7 μM and 2 μM, respectively. NG 52 also inhibits the activity of phosphoglycerate kinase 1 (PGK1) with an IC50 of 2.5 μM. NG 52 is inactive against yeast kinases Kin28p, Srb10, and Cak1p.

NG 52 is a potent, cell-permeable, selective, ATP-compatible and orally active Cdc28p and Pho85p kinase inhibitor with IC50s of 7 μM and 2 μM, respectively. NG 52 also inhibits the activity of phosphoglycerate kinase 1 (PGK1) with an IC50 of 2.5 μM. NG 52 is inactive against yeast kinases Kin28p, Srb10, and Cak1p.

Specifications

Chemical Name or Material NG 52
CAS 212779-48-1
Color White
Purity Grade Notes Research
Molecular Formula C16H19ClN6O
Quantity 50 mg
Synonym Compound 52
Solubility Information DMSO : 100 mg/mL (288.34 mM; Need ultrasonic)
SMILES ClC1=CC(NC2=NC(NCCO)=NC3=C2N=CN3C(C)C)=CC=C1
Molecular Weight (g/mol) 346.81
Formula Weight 346.81
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.