missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NG 25
GREENER_CHOICE

Product Code. 18723439
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18723439 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18723439 Supplier Tocris Bioscience™ Supplier No. 5429/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TGF-β-activated kinase (TAK1) inhibitor

NG 25 is a TGF-β-activated kinase (TAK1) inhibitor (IC50 = 149 nM). Also inhibits LYN, MAP4K2 and Abl kinase activity (IC50 values are 12.9, 21.7 and 75.2 nM, respectively). Suppresses CpG A and CpG B-stimulated secretion of IFN-α and TLR7/8 agonist CL097-stimulated secretion of IFN-β in Gen 2.2 leukemia cells in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-[(4-Ethyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-4-methyl-3-(1H-pyrrolo[2,3-b]pyridin-4-yloxy)benzamide
CAS 1315355-93-1
Quantity 5 mg
Target Other Kinase Inhibitors
Molecular Formula C29H30F3N5O2
Formula Weight 537.58
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.