missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFYC, Mouse anti-Human, Clone: 1D3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004802-M01.50ug
This item is not returnable.
View return policy
Description
Sequence: DAQQSLQSFWPRVMEEIRNLTVKDFRVQELPLARIKKIMKLDEDVKMISAEAPVLFAKAAQIFITELTLRAWIHTEDNKRRTLQRNDIAMAITKFDQFDFSpecifications
| NFYC | |
| Monoclonal | |
| Unconjugated | |
| nuclear transcription factor Y, gamma | |
| CBF-C/CBFC/DKFZp667G242/FLJ45775/H1TF2A/HAP5/HSM/NF-YC | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown, Western Blot | |
| 1D3 | |
| In 1x PBS, pH 7.4 | |
| BC005003 | |
| NFYC | |
| NFYC (AAH05003, 14 a.a. ∼ 113 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4802 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction