missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NFX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566383
This item is not returnable.
View return policy
Description
Immunogen sequence: VFHPDSSEASS RKGVLDGYGA RRNEQRRYPQ KRPPWEVEGA RPRPGRNPPK QEGHRHTNAG HRNNMGPIPK DDLNERPAKS TCDSENLAVI NKSSR Highest antigen sequence identity to the following orthologs - mouse 73%, rat 73%.
MHC class II gene expression is controlled primarily at the transcriptional level by transcription factors that bind to the X and Y boxes, two highly conserved elements in the proximal promoter of MHC class II genes. NFX1 is a transcriptional repressor capable of binding to the conserved X box motif of HLA-DRA and other MHC class II genes in vitro. The protein may play a role in regulating the duration of an inflammatory response by limiting the period in which class II MHC molecules are induced by IFN-gamma.MHC class II gene expression is controlled primarily at the transcriptional level by transcription factors that bind to the X and Y boxes, two highly conserved elements in the proximal promoter of MHC class II genes. The protein encoded by this gene is a transcriptional repressor capable of binding to the conserved X box motif of HLA-DRA and other MHC class II genes in vitro. The protein may play a role in regulating the duration of an inflammatory response by limiting the period in which class II MHC molecules are induced by IFN-gamma. Three alternative splice variants, each of which encodes a different isoform, have been identified.
Specifications
| NFX1 | |
| Polyclonal | |
| Unconjugated | |
| NFX1 | |
| 1300017N15Rik; 3000003M19Rik; DKFZp779G2416; MGC20369; m-Nfx.1; NFX.1; Nfx1; NFX2; nuclear transcription factor, X box-binding protein 1; nuclear transcription factor, X-box binding 1; TEG-42; testis expressed gene 42; Tex42; transcriptional repressor NF-X1; X-box binding 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4799 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q12986 | |
| NFX1 | |
| Recombinant Human NFX1. Recombinant protein control fragment (Product #RP-105180). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction