missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFX1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49932PCP
This item is not returnable.
View return policy
Description
NFX1 Polyclonal antibody specifically detects NFX1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Specifications
| NFX1 | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| DKFZp779G2416, EC 6.3.2.-, MGC20369, NFX2transcriptional repressor NF-X1, Nuclear transcription factor, X box-binding protein 1, nuclear transcription factor, X-box binding 1 | |
| The immunogen for this product maps to a region between residue 250 and 300 of human Nuclear Transcription Factor, X-box Binding 1 using the numbering given in entry NP_002495.2 (GeneID 4799). | |
| 0.1 mL | |
| Zinc Finger | |
| 4799 | |
| Store at 4C in the dark. | |
| IgG |
| Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction