missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFRKB Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£145.00 - £383.00
Specifications
| Antigen | NFRKB |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232389
|
Novus Biologicals
NBP3-33460-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227589
|
Novus Biologicals
NBP3-33460-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NFRKB Monoclonal antibody specifically detects NFRKB in Human samples. It is validated for ELISA,Western BlotSpecifications
| NFRKB | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp547B2013, DNA-binding protein R kappa-B, INO80 complex subunit G, INO80GDNA-binding protein R kappa B, nuclear factor related to kappaB binding protein, nuclear factor related to kappa-B-binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 370-495 of human NFRKB (NP_001137307.1).,, Sequence:, LGINEISSSFFSLLLEILLLESQASLPMLEERVLDWQSSPASSLNSWFSAAPNWAELVLPALQYLAGESRAVPSSFSPFVEFKEKTQQWKLLGQSQDNEKELAALFQLWLETKDQAFCKQENEDSS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 4798 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title