missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFRKB Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | NFRKB |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
NFRKB Polyclonal antibody specifically detects NFRKB in Human samples. It is validated for Western BlotSpecifications
| NFRKB | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp547B2013, DNA-binding protein R kappa-B, INO80 complex subunit G, INO80GDNA-binding protein R kappa B, nuclear factor related to kappaB binding protein, nuclear factor related to kappa-B-binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-340 of human NFRKB (NP_001137307.1). MDSLDHMLTDPLELGPCGDGHGTRIMEDCLLGGTRVSLPEDLLEDPEIFFDVVSLSTWQEVLSDSQREHLQQFLPQFPEDSAEQQNELILALFSGENFRFGNPLHIAQKLFRDGHFNPEVVKYRQLCFKSQYKRYLNSQQQYFHRLLKQILASRSDLLEMARRSGPALPFRQKRPSPSRTPEEREWRTQQRYLKVLREVKEECGDTALSSDEEDLSSWLPSSPARSPSPAVPLRVVPTLSTTDMKTADKVELGDSDLKIMLKKHHEKRKHQPDHPDLLTGDLTLNDIMTRVNAGRKGSLAALYDLAVLKKKVKEKEEKKKKKIKTIKSEAEDLAEPLSST | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 4798 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title