missing translation for 'onlineSavingsMsg'
Learn More

NFKBIL1 Antibody - BSA Free, Novus Biologicals™

Product Code. 18661081 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18661081 0.02 mL 0.02mL
18696941 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18661081 Supplier Novus Biologicals Supplier No. NBP2935320.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NFKBIL1 Polyclonal antibody specifically detects NFKBIL1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFKBIL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias IkappaBL, I-kappa-B-like protein, IKBLikappaBL, Inhibitor of kappa B-like protein, LST1, NF-kappa-B inhibitor-like protein 1, nuclear factor of kappa light polypeptide gene enhancer in B-cellsinhibitor-like 1NFKBIL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-381 of human NFKBIL1 (NP_004998.3). MSNPSPQVPEEEASTSVCRPKSSMASTSRRQRRERRFRRYLSAGRLVRAQALLQRHPGLDVDAGQPPPLHRACARHDAPALCLLLRLGADPAHQDRHGDTALHAAARQGPDAYTDFFLPLLSRCPSAMGIKNKDGETPGQILGWGPPWDSAEEEEEDDASKEREWRQKLQGELEDEWQEVMGRFEGDASHETQEPESFSAWSDRLAREHAQKCQQQQREAEGSRRPPRAEGSSQSWRQQEEEQRLFRERARAKEEELRESRARRAQEALGDREPKPTRAGPREEHPRGAGRGSLWRFGDVPWPCPGGGDPEAMAAALVARGPPLEEQGALRRYLRVQQVRWHPDRFLQRFRSQIETWELGRVMGAVTALSQALNRHAEALK
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4795
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.