missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFKBIE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11273-1-AP-150UL
This item is not returnable.
View return policy
Description
Inhibits NF-kappa-B by complexing with and trapping it in the cytoplasm. Inhibits DNA-binding of NF-kappa-B p50-p65 and p50-c-Rel complexes.Specifications
| NFKBIE | |
| Polyclonal | |
| Unconjugated | |
| NFKBIE | |
| I kappa B epsilon, IkappaBepsilon, IkB E, IkB epsilon, IKBE, NF kappa B inhibitor epsilon, NF kappa BIE, NFKBIE | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18037, 316241, 4794 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00221, O54910 | |
| Nfkbie | |
| NFKBIE Fusion Protein Ag1800 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction